CO1H324A62A2 CombiPhos Catalysts Inc

(2-ACETYLPYRIMIDIN-5-YL)BORONIC ACID

  • Cas Number : 1229672-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart