CO1H324A62A5 CombiPhos Catalysts Inc

(4-(azetidin-3-yl)phenyl)boronic acid

  • Cas Number : 1906861-55-9
  • Molecular Weight : 177.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart