CO1H324A62A7 CombiPhos Catalysts Inc

(3-fluoro-4-(oxetan-3-yl)phenyl)boronic acid

  • Cas Number : 2256708-82-2
  • Molecular Weight : 195.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart