CO1H324A62B2 CombiPhos Catalysts Inc

(2-(pyridin-2-yl)pyrimidin-5-yl)boronic acid

  • Cas Number : 2225172-14-3
  • Molecular Weight : 200.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart