CO1H324A62B5 CombiPhos Catalysts Inc

(4-(pyrrolidin-3-yl)-3-(trifluoromethyl)phenyl)boronic acid

  • Cas Number : 2256708-86-6
  • Molecular Weight : 259.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart