CO1H324A62B6 CombiPhos Catalysts Inc

(4-(piperidin-1-yl)-3-(trifluoromethyl)phenyl)boronic acid

  • Cas Number : 1187874-86-7
  • Molecular Weight : 273.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart