CO1H324A62C1 CombiPhos Catalysts Inc

1-(4-Pyridyl)-1H-pyrazole-4-boronic acid

  • Cas Number : 1802481-47-5
  • Molecular Weight : 188.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart