CO1H324A62D0 CombiPhos Catalysts Inc

1-(3-Methoxyphenyl)-1H-pyrazole-4-boronic acid

  • Cas Number : 2225177-37-5
  • Molecular Weight : 218.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart