CO1H324A62D8 CombiPhos Catalysts Inc

1-(4-Cyanophenyl)-1H-pyrazole-4-boronic acid

  • Cas Number : 1802481-33-9
  • Molecular Weight : 213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart