CO1H324A62E1 CombiPhos Catalysts Inc

1-(4-tert-Butylphenyl)-1H-pyrazole-4-boronic acid

  • Cas Number : 2225169-39-9
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart