CO1H324A62E2 CombiPhos Catalysts Inc

1-(2-Chloro-4-pyridyl)-1H-pyrazole-4-boronic acid

  • Cas Number : 2225181-14-4
  • Molecular Weight : 223.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart