CO1H324A62EA CombiPhos Catalysts Inc

3-Methyl-1-(thiazol-5-yl)pyrazole-4-boronic acid

  • Cas Number : 2225177-44-4
  • Molecular Weight : 209.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart