CO1H324A62F0 CombiPhos Catalysts Inc

3-Methyl-1-(pyridin-3-yl)pyrazole-4-boronic acid

  • Cas Number : 1394837-47-8
  • Molecular Weight : 203.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart