CO1H324A62F5 CombiPhos Catalysts Inc

3-Methyl-1-(3-furyl)pyrazole-4-boronic acid

  • Cas Number : 2225178-86-7
  • Molecular Weight : 191.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart