CO1H324A62F7 CombiPhos Catalysts Inc

3-Methyl-1-(2-fluorophenyl)pyrazole-4-boronic acid

  • Cas Number : 2225177-54-6
  • Molecular Weight : 220.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart