CO1H324A6301 CombiPhos Catalysts Inc

4-Chloro-2-ethylpyrimidine-5-boronic acid

  • Cas Number : 2225169-99-1
  • Molecular Weight : 186.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart