CO1H324A631A CombiPhos Catalysts Inc

4-Chloro-2-(2-trifluoromethylphenyl)pyrimidine-5-boronic acid

  • Cas Number : 2225170-02-3
  • Molecular Weight : 302.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart