CO1H324A6329 CombiPhos Catalysts Inc

(2-(methoxycarbonyl)cyclopropyl)boronic acid

  • Cas Number : 1422542-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart