CO1H324A632A CombiPhos Catalysts Inc

2-(Cyclopropanecarbonyl)cyclopropane boronic acid

  • Cas Number : 2225152-57-6
  • Molecular Weight : 153.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart