CO1H324A632B CombiPhos Catalysts Inc

2-Cyano-2-(methoxycarbonyl)cyclopropyl boronic acid

  • Cas Number : 2225152-96-3
  • Molecular Weight : 168.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart