CO1H324A6332 CombiPhos Catalysts Inc

2,2-Bis(ethoxycarbonyl)cyclopropyl boronic acid

  • Cas Number : 2225152-08-7
  • Molecular Weight : 230.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart