CO1H324A6339 CombiPhos Catalysts Inc

(5-Methyl)tetrahydrofuran-3- boronic acid

  • Cas Number : 2225152-41-8
  • Molecular Weight : 129.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart