CO1H324A633B CombiPhos Catalysts Inc

2-Chloro-4-methylpyrimidine-5-boronic acid

  • Cas Number : 2225155-62-2
  • Molecular Weight : 172.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart