CO1H324A633F CombiPhos Catalysts Inc

(6-(thiazol-2-yl)pyridin-3-yl)boronic acid

  • Cas Number : 1264511-30-9
  • Molecular Weight : 206.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart