CO1H324A6342 CombiPhos Catalysts Inc

4-Cyclopropylpyrimidine-5-boronic acid pinacol ester

  • Cas Number : 1375303-49-3
  • Molecular Weight : 246.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart