CO1H324A6343 CombiPhos Catalysts Inc

4-Aminopyrimidine-5-boronic acid pinacol ester

  • Cas Number : 2223044-20-8
  • Molecular Weight : 221.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart