CO1H324A6349 CombiPhos Catalysts Inc

4-Amino-2-(cyclobutyll)pyrimidine-5-boronic acid pinacol ester

  • Cas Number : 2223031-18-1
  • Molecular Weight : 275.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart