CO1H324A634D CombiPhos Catalysts Inc

4-Amino-2-(pyridin-4-yl)pyrimidine-5-boronic acid pinacol ester

  • Cas Number : 2223031-74-9
  • Molecular Weight : 298.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart