CO1H324A6357 CombiPhos Catalysts Inc

2-Amino-4-(iso-propyl)pyrimidine-5-boronic acid pinacol ester

  • Cas Number : 2223041-79-8
  • Molecular Weight : 263.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart