CO1H324A6359 CombiPhos Catalysts Inc

3-Aminopyridazine-4-boronic acid pinacol ester

  • Cas Number : 2223039-24-3
  • Molecular Weight : 221.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart