CO1H324A6360 CombiPhos Catalysts Inc

4-(4-Fluorophenyl)thiazole-5-boronic acid pinacol ester

  • Cas Number : 2223044-42-4
  • Molecular Weight : 305.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart