CO1H324A6365 CombiPhos Catalysts Inc

4-(3-Trifluoromethylphenyl)thiazole-5-boronic acid pinacol ester

  • Cas Number : 2223031-87-4
  • Molecular Weight : 355.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart