CO1H324A636C CombiPhos Catalysts Inc

5-(tert-Butyl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 1251848-80-2
  • Molecular Weight : 266.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart