CO1H324A636D CombiPhos Catalysts Inc

5-Cyclohexylthiophene-2-boronic acid pinacol ester

  • Cas Number : 1402166-55-5
  • Molecular Weight : 292.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart