CO1H324A636E CombiPhos Catalysts Inc

5-(3-Thienyl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 1402166-53-3
  • Molecular Weight : 292.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart