CO1H324A6375 CombiPhos Catalysts Inc

5-(4-Pyridyl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 1402240-70-3
  • Molecular Weight : 287.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart