CO1H324A637A CombiPhos Catalysts Inc

5-(4-Trifluoromethylphenyl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 1402234-05-2
  • Molecular Weight : 354.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart