CO1H324A6383 CombiPhos Catalysts Inc

5-(2-isopropyl-5-pyrimidyl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 1402233-33-3
  • Molecular Weight : 330.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart