CO1H324A6387 CombiPhos Catalysts Inc

4-(3,4-Difluorophenyl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 1402227-89-7
  • Molecular Weight : 322.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart