CO1H324A6394 CombiPhos Catalysts Inc

4-(4-Chlorophenyl)thiophene-2-boronic acid pinacol ester

  • Cas Number : 1402227-79-5
  • Molecular Weight : 320.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart