CO1H324A6396 CombiPhos Catalysts Inc

2-(N-diphenylmethylene)thiazole-5-boronic acid pinacol ester

  • Cas Number : 2223041-68-5
  • Molecular Weight : 390.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart