CO1H324A639B CombiPhos Catalysts Inc

5-(n-Propyl)thiazole-4-boronic acid pinacol ester

  • Cas Number : 2223039-22-1
  • Molecular Weight : 253.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart