CO1H324A639D CombiPhos Catalysts Inc

5-(Cyclopropyl)thiazole-4-boronic acid pinacol ester

  • Cas Number : 2223031-37-4
  • Molecular Weight : 251.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart