CO1H324A63A3 CombiPhos Catalysts Inc

5-(3-Tolyl)thiazole-4-boronic acid pinacol ester

  • Cas Number : 2223011-17-2
  • Molecular Weight : 301.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart