CO1H324A63A4 CombiPhos Catalysts Inc

5-(4-Tolyl)thiazole-4-boronic acid pinacol ester

  • Cas Number : 2223044-52-6
  • Molecular Weight : 301.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart