CO1H324A63A7 CombiPhos Catalysts Inc

5-(3-Methoxyphenyl)thiazole-4-boronic acid pinacol ester

  • Cas Number : 2223050-75-5
  • Molecular Weight : 317.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart