CO1H324A63A9 CombiPhos Catalysts Inc

5-(2-Fluorophenyl)thiazole-4-boronic acid pinacol ester

  • Cas Number : 2223033-23-4
  • Molecular Weight : 305.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart