CO1H324A63B0 CombiPhos Catalysts Inc

5-(3,5-Difluorophenyl)thiazole-4-boronic acid pinacol ester

  • Cas Number : 2223039-36-7
  • Molecular Weight : 323.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart