CO1H324A63B3 CombiPhos Catalysts Inc

5-(1-Hydroxy-1-methylethyl)furan-2-boronic acid pinacol ester

  • Cas Number : 2223030-67-7
  • Molecular Weight : 252.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart