CO1H324A63B9 CombiPhos Catalysts Inc

2-((tetrahydrofuran-3-yl)oxy)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazole

  • Cas Number : 1356369-13-5
  • Molecular Weight : 297.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart